Cleanroom wipes, pre-saturated, AlphaSat®
Supplier: Texwipe
TX1034 TX1036 TX1039
115-0456EA 234 GBP
115-0456 115-0457 115-0458
Cleanroom wipes, pre-saturated, AlphaSat®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These highly sorbent wipes are made from 100% continuous filament polyester in a double-knit, 'no run' interlock pattern. The wipes are pre-wetted with 70% IPA and 30% deionised water.
  • Superior strength with low particle generation and low extractable levels
  • Laundered cut edge
  • Excellent chemical and abrasion resistance
Ideal for general wiping, spill control, polishing and cleaning in critical environments (ISO Class 4 to 9).
Colour: White
Packaging: Solvent safe, reclosable packaging.
Caution: Data accurate at time of publishing, latest testMore Product Information