ESD safety shoes, lace-up, uvex 1 G2 planet, 6823
Supplier: UVEX
68232/36 68232/37 68232/38 68232/39 68232/41 68232/42 68232/43 68232/44 68232/45 68232/46 68232/47
114-9105EA 0 GBP
114-9105 114-9106 114-9107 114-9108 114-9110 114-9111 114-9112 114-9113 114-9114 114-9115 114-9116
ESD safety shoes, lace-up, uvex 1 G2 planet, 6823
Shoes and Boots
These extremely lightweight S1 P safety shoes are made from recycled and sustainable materials.
  • Made from synthetic materials, making it suitable for people allergic to chrome
  • Meets the ESD specification with volume resistance under 35 megaohms
  • Elastic laces for quick, individual adjustment (normal laces also provided)
  • Soft padding on dust tongue and collar
  • Virtually seam-free upper design made from textile material to prevent pressure points
  • uvex x-dry knit planet textile material made from 100% recycled PET bottles
  • New uvex xenova® composite toe cap and
More Product Information