Aluminium 99,997 Metals basis, foil, Puratronic®, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminum

11375.GH 11375.HP
11375.GHEA 95.4 GBP
11375.GH 11375.HP
Aluminium 99,997 Metals basis, foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Aluminium

Purity: 99.997% CAS No.: 7429-90-5 Physical Form: ≈1.7g/50x50mm, Temper: as rolled Odour: Odourless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR