BioClean-C™ Chemo Protective Aprons with Sleeves
Supplier: Nitritex
BCAS-L BCAS-M BCAS-S S-BCAS-L S-BCAS-M S-BCAS-S BCAS-XL
113-8146EA 240 GBP
113-8146 113-8147 113-8148 113-8162 113-8163 113-8164 NITRBCAS-XL
BioClean-C™ Chemo Protective Aprons with Sleeves
Disposable Garments
These disposable aprons are made of CleanTough™ fabric, a spunbonded non woven PP fabric laminated with a PE film. The elasticated cuffs are made of 100% polyester.
  • Processed to ensure ISO Class 4 compatibility
  • Lightweight, low linting and hard-wearing CleanTough™ fabric
  • Ultrasonically bonded seams with protective tape
  • Adjustable neck fastening
  • Tie tapes at the waist
Provide protection against fine sprays and particles as well as against a range of chemotherapy drugs.
Colour: Blue.
Certifications: EN ISO 13034:2005 + A1:2010 (Type PB [6]),More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Fabric CleanTough™ (PP/PE)
Cleanroom class 10/ISO 4
Gender
Sleeve or leg length Long
Garment application Cleanroom,Medical,Pharmaceutical,Clinical