Laboratory coat, KIMTECH SCIENCE* A7 P+
Supplier: KIMBERLY CLARK
97740 97700 97710 97730 97720
113-7901EA 85.9 GBP
113-7901 113-7897 113-7898 113-7900 113-7899
Laboratory coat, KIMTECH SCIENCE* A7 P+
Disposable Garments
These lab coats made from film coated PP with triple stitched seams are ideal for use in both research and manufacturing environments in pharmaceutical, biochemistry, biomedical, cosmetics, forensics, and other applications. They protect the user against low risk liquid chemicals in the form of light splashes, sprays and aerosols.
  • High levels of particle and bacterial protection (with filtration efficiency of 99%)
  • Non woven material has a cloth-like feel and is breathable, reducing the risk of heat stress and making the lab coats suitable for long periods
More Product Information