Shoe covers, non skid, VWR® Advanced, SF
Supplier: VWR Collection
113-1383EA 80.4 GBP
113-1383 113-1382 113-1378 113-1377 113-1384
Shoe covers, non skid, VWR® Advanced, SF
Disposable Garments
VWR® Advanced laminated shoe covers are manufactured from spunbonded polypropylene fabric (SPP), coated with polyethylene film.
  • High coefficient of friction to provide excellent traction
  • Water-resistant
  • Durable fabric with good breathability
  • Significant fluid and particulate barrier
  • Particle shedding: Level II Helmke drum classification
  • Soft, cloth-like fabric
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Length: 394 mm (U*), 432 mm (XL), 47More Product Information