Cleanroom lab coats, BioClean-D™, CleanTough™, disposable
Supplier: Nitritex
BDLC-L BDLC-M BDLC-S BDLC-XL BDLC-XXL S-BDLC-M S-BDLC-S S-BDLC-XL S-BDLC-XXL S-BDLC-L
113-0809EA 222 GBP
113-0809 113-0810 113-0811 113-0812 113-0813 113-0825 113-0826 113-0827 113-0828 113-0824
Cleanroom lab coats, BioClean-D™, CleanTough™, disposable
Disposable Garments
These disposable lab coats are made from CleanTough™ fabric which is spunbonded non-woven PP laminated with a film of PE. The fabric provides protection against fine sprays and particles.
  • Lightweight, low linting, and hard-wearing material
  • Three pockets, press stud fastening
  • Open cuffs and rear vent
  • Available as sterile or non sterile version
  • A unique barcode on the outer carton (ITF-14) allows automatic data entry into Stock Control Systems
Suitable for use in Class 10, ISO 4, M2.5 environments.
Colour: White
Type PB [6]; Cat. III