General purpose gloves, Showa 7199NC
Supplier: Showa
7199NC
112-5893EA 951 GBP
112-5893
General purpose gloves, Showa 7199NC
Gloves
This flexible cotton jersey liner with full nitrile coating provides good mechanical resistance. Recommended for heavy-duty risks.
  • Absorbs perspiration, for comfortable extended wear
  • Protects the hand from oils and abrasion while retaining good touch
  • Impermeable for working in damp or greasy environments
  • Antibacterial and anti-odour treatment
  • Topstitched edge
Ideal for agriculture, automotive industry, food applications, glass and metallurgy, petrochemical industry and the public sector.
Certifications: EN 388:2016 (4111X); Cat. II.
Order Now
SPECIFICATIONS
Model 7199NC
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable
Liner material Jersey
Finish Smooth
EN norm EN 388:2016 (4111X)