Cut resistant gloves, Aracut® Lat
Supplier: HONEYWELL SAFETY
2094141/10 2094141/7 2094141/8 2094141/9
112-1609EA 0 GBP
112-1609 112-1610 112-1611 112-1612
Cut resistant gloves, Aracut® Lat
Gloves
Knitted gloves made from 100% Kevlar® with knitted cuffs and reinforced palms and fingertips, made from blue creped latex.
  • Natural latex coating offers excellent flexibility and a secure grip
  • Optimum breathability due to ventilated back
  • Creped latex coating ensures good mechanical resistance to perforation and abrasion
  • Additional coating between the thumb and forefinger reinforces the wear areas and increases the lifespan of the glove
Ideal for handling sharp parts in a dry or damp environment.
Gauge: 10
Colour: blue/beige
EN 388 (3344), Cat.More Product Information
Order Now
SPECIFICATIONS
Model Lat
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable
Liner material Kevlar®
Finish Coated
Gauge 10
Cuff style Knit wrist
EN norm EN 388:2016 (3X44C), EN 407 (X2XXXX)