Battery, Versaflo™ TR-830
Supplier: 3M
TR-830
111-2548EA 1040 GBP
111-2548
Battery, Versaflo™ TR-830
Batteries
The fast-charging, intrinsically safe Versaflo™ lithium-ion battery TR 830 is intended for use with the 3M™ Versaflo™ TR 800 series PAPR system.
The battery reaches 90% charge in approximately three and a half hours and features a charge level indicator for easy visual reference. It is engineered for use in potentially explosive athmospheres and features yellow bumpers to help absorb impact.
When equipped with a new intrinsically safe battery (TR-830) and new filter, the TR-800 PAPR assembly has a run time of 6 to 10 hours depending on system configuration used. PAPR runtime will varyMore Product Information