Safety helmets, uvex pheos E-S-WR
Supplier: UVEX
9770.131 9770.832 9770.031 9770.431
111-2335EA 22.6 GBP
111-2335 111-2336 111-2334 UVEX9770.431
Safety helmets, uvex pheos E-S-WR
Head Protection Safety Helmets
The lightweight uvex pheos safety helmets feature a sporty design and offer maximum wearer comfort. The sturdy, rigid helmet shell also easily withstands pressure from the side, and fulfills all safety related requirements for construction or industrial work.
  • Can quickly and easily be adapted to the user's individual needs - visor, safety spectacles, ear defenders or head torch
  • Suspension harness with wheel ratchet for variable width adjustment
  • Six-point textile band attachment ensures optimal fit and comfort
  • With slot on the front for helmet lights and
More Product Information