Escape hoods, Parat® 7500
Supplier: Dräger
R59437
111-2171EA 351 GBP
111-2171
Escape hoods, Parat® 7500
Respirators Respiratory Systems
This combined fire and industrial escape hood was developed with users, placing the focus on the fastest possible escape. Optimised operation and wearing comfort, a robust housing and a tested ABEK CO P3 filter ensure protection from toxic industrial and fire-related gases, vapours and particles for at least 15 minutes.
  • Hood in signal colour for high visibility
  • Automatically adjusting internal head harness: No additional tighting of the hood is required
  • Flexible textile neck seal
  • Exhalation valve directly leads the exhaled air out of the half mask
  • Wide
More Product Information