Safety spectacles, SecureFit™ 200
Supplier: 3M
SF202AF SF203AF
111-1593EA 0 GBP
111-1593 111-1594
Safety spectacles, SecureFit™ 200
Safety Glasses
Stylish, lightweight designed safety glasses features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
  • 3M Pressure Diffusion Technology allows temples to naturally adjust to individual head sizes
  • Self-adjusting feature eliminates need to stock multiple frame sizes at worksites
  • Secure, snug fit keeps eyewear in place without slippage even when the person is in motion
Weight: 18 g
Certifications: EN 166.