Disposable particle filtering respirators, FFP1/FFP2/FFP3, X-plore® 1700
Supplier: Dräger
3951086 3951088 3951080 3951085 3951083 3951081 3951084 3951196
111-1037EA 154 GBP
111-1037 111-1038 111-0756 111-0758 111-1036 111-1034 111-0757 111-2517
Disposable particle filtering respirators, FFP1/FFP2/FFP3, X-plore® 1700
Respirators Disposable Respirators Particulate Respirators
The X-plore® 1700 series offers models in all three EN protection classes: FFP1, FFP2, and FFP3. Available with or without CoolMax™ exhalation valve for reduced exhalation resistance and to avoid a build-up of heat. Odour models for tackling unpleasant odours are equipped with an additional activated carbon layer.
  • Special high performance filter media are brought together to form the CoolSafe™ filter material, ensuring a low level of exhalation resistance for users while delivering a high level of filter performance
  • Elasticated VarioFlex™ head strap to
More Product Information