Earmuffs, Peltor™ Optime™ I
Supplier: 3M
H510A H510F H510B H510P3E
111-0673EA 30.7 GBP
111-0673 111-0674 111-1075 111-0676
Earmuffs, Peltor™ Optime™ I
Earmuffs Noise Blocking Earmuffs
The lightweight Peltor™ Optime™ I earmuffs provide a moderate level of attenuation that meets the needs of the majority of industrial applications.
  • Liquid-sealing rings for improved comfort
  • Large space inside cups helps reduce moisture and heat build-up
  • Soft, wide cushions reduce pressure around the ears
  • Easy to replace cushions and inserts help keep them hygienically clean
  • Helmet-mounted version fits directly to many safety helmets without the need for an adapter
For industrial applications, such as workshops, sheet metal shops and printing works.More Product Information