Potassium carbonate sesquihydrate 98,5-101,0 ACS

Supplier: Thermo Fisher Scientific
Warning

10981.22 10981.36
10981.22EA 33.6 GBP
10981.22 10981.36
Potassium carbonate sesquihydrate 98,5-101,0 ACS
Potassium carbonate sesquihydrate

Purity: 98.5-101.0% CAS No.: 6381-79-9 Molecular Formula: K2CO3.1.5H2O Formula Weight: 165.23 (138.21anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Solubility: Soluble in water. Practically insoluble in alcohol Merck Reference: 14,7670 MDL No.: MFCD00149925


Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent

Formula: CK₂O₃·1,5H₂O
MW: 165.23 g/mol
Melting Pt: 891
Density: 2.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149925
CAS Number: 6381-79-9
EINECS: 209-529-3
Merck Index: 14,7670

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR