Nickel (II) oxide ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

10819.06 10819.14 10819.22
10819.06EA 47.2 GBP
10819.06 10819.14 10819.22
Nickel (II) oxide ≥99.998% (metals basis), Puratronic®
Nickel (II) oxide

Purity: 99.998% CAS No.: 1313-99-1 Molecular Formula: NiO Formula Weight: 74.71 Physical Form: Powder Density: 6.67 Appearance: Dark green Odour: Odourless Melting Point: 1984° Merck Reference: 14,6512 MDL No.: MFCD00011145

Formula: NiO
MW: 74.69 g/mol
Melting Pt: 1984 °C
Density: 6.67
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7
Merck Index: 14,6512

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR