DNA transfection kit for protein production, FectoCHO® Expression system
Supplier: Polyplus-transfection
101000003 101000011
101000003.EA 1070 GBP
101000003. 101000011.
DNA transfection kit for protein production, FectoCHO® Expression system
Transfection Reagents
FectoCHO® expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD expression medium and a powerful transfection reagent, FectoPRO®.
  • Outstanding protein production yields in various CHO cells
  • Fast and effortless CHO cell adaptation
  • Chemically defined kit components
  • Excellent lot to lot reproducibility
  • Cost-effective solution
FectoCHO® CD expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with similar systems.
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Ordering information: FectoPRO® transfection reagent and FectoCHO® CD expression medium can be purchased separately.
Order Now
SPECIFICATIONS
Cell specific HEK-293, CHO
Environmentally Preferable
Bioproduction Yes
DNA Yes
DNA and siRNA Yes
siRNA Yes