Manganese standard solution, 10000 mg/L Mn in dil. nitric acid (from Mn(NO₃)₂) Certipur® standard for ICP, Supelco®
Supplier: Merck
1.70380.0100
1.70380.0100EA 236 GBP
1.70380.0100
Manganese standard solution, 10000 mg/L Mn in dil. nitric acid (from Mn(NO₃)₂) Certipur® standard for ICP, Supelco®
Standards Cationic Standards Manganese Standards
Certifications: Traceable to SRM from NIST