, LiChrosolv®, hypergrade LC-MS, Supelco®

Supplier: Merck
Danger


MQ100

1.59002.1000 1.59002.2500 1.59002.4000
1.59002.1000EA 115 GBP
1.59002.1000 1.59002.2500 1.59002.4000
, LiChrosolv®, hypergrade LC-MS, Supelco®
Acetonitrile with 0,1% (v/v) Formic acid

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to prefectly meet LC-MS quality requirements.

Boiling Pt: 81.6 °C (1013 hPa)
Flash Pt: 2 °C
Storage Temperature: Ambient
UN: 1648
ADR: 3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR