tri-Sodium citrate dihydrate, EMPROVE® EXPERT USP, BP, Ph. Eur., ACS, JP BiopharmaceuticalProduction, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

1.37042.5000 1.37042.9012 1.37042.9029
1.37042.5000EA 757 GBP
1.37042.5000 1.37042.9012 1.37042.9029
tri-Sodium citrate dihydrate, EMPROVE® EXPERT USP, BP, Ph. Eur., ACS, JP BiopharmaceuticalProduction, SAFC®
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR