Phenol red, EMPROVE® ESSENTIAL, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

1.37038.0100
1.37038.0100EA 0 GBP
1.37038.0100
Phenol red, EMPROVE® ESSENTIAL, SAFC®
Phenol red
Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Merck Index: 13,07329

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR