System for testing jet fuel, HY-LiTE® Jet A1 Fuel Test
Supplier: Merck
1.30196.0021
1.30196.0021EA 1330 GBP
1.30196.0021
System for testing jet fuel, HY-LiTE® Jet A1 Fuel Test
Monitoring Systems Hygiene Monitoring Systems
For aviation fuel used in civilian air transport. Aviation fuel contains small quantities of water, so it is vulnerable to microbiological attack. Microbiological contamination in fuel can lead to operational problems, e.g. clogged filters or corroded tanks. This test is an alternative for anyone who monitors or deals with aviation fuel. Previously, test results were only available after three days. With the HY-LiTE® test, results are returned in just 10 minutes. This test is recommended in the IATA (International Air Transport Association) Directive.
  • A simple, on-site test,
More Product Information