Test strips, peracetic acid, Reflectoquant®, Supelco®
Supplier: Merck
1.16975.0001 1.16976.0001 1.17956.0001
1.16975.0001EA 47.6 GBP
1.16975.0001 1.16976.0001 1.17956.0001
Test strips, peracetic acid, Reflectoquant®, Supelco®
Environmental Test Kits
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Applications: Disinfectant solutions - in cases in which hydrogen peroxide is present.
Order Now
SPECIFICATIONS
Environmentally Preferable
M-Clarity quality level MQ100