Test strips, formaldehyde, Reflectoquant®, Supelco®
Supplier: Merck
1.16989.0001
1.16989.0001EA 156 GBP
1.16989.0001
Test strips, formaldehyde, Reflectoquant®, Supelco®
Environmental Test Kits
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Order Now
SPECIFICATIONS
Environmentally Preferable
M-Clarity quality level MQ100