Germanium(IV) oxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Germanium dioxide

MQ200

1.12177.0005 1.12177.0025 1.12177.0002
1.12177.0005EA 87.8 GBP
1.12177.0005 1.12177.0025 1.12177.0002
Germanium(IV) oxide, Sigma-Aldrich®
Germanium (IV) oxide
Formula: GeO₂
MW: 104.61 g/mol
Melting Pt: 1110
Density: 3.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417

Order Now

Specification Test Results

Solubility 4.5 g/l (25 °C)
Melting Point 1115 °C
Molar Mass 104.61 g/mol
Bulk Density 1430 kg/m3
Density 4.23 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR