Pyridine, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Supplier: Merck
Danger

Synonyms: Azabenzene, Azine

MQ300

1.09728.0100 1.09728.0500 1.09728.1000 1.09728.2500 1.09728.6025 1.09728.4000
1.09728.0100EA 43.2 GBP
1.09728.0100 1.09728.0500 1.09728.1000 1.09728.2500 1.09728.6025 1.09728.4000
Pyridine, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
Pyridine

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -42 °C
Molar Mass 79.1 g/mol
Boiling Point 115 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Flash Point 17 °C
Explosion Limit 1.7 - 10.6 %(V)
Density 0.982 g/cm3 (20 °C)
pH 8.5 (16 g/l, H2O, 20 °C)
Ignition Point 482 °C
Evaporation number 12.7

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR