Buffer solutions, Certipur®, colour coded, Supelco®

Supplier: Merck

1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
1.09475.0500EA 29.96 GBP
1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
Buffer solutions, Certipur®, colour coded, Supelco®
Buffers pH Calibration Buffers

Materials for the precise calibration and monitoring of pH meters.


  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C


Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR