Air sampler, MAS-100 NT® Ex
Supplier: Merck
1.09194.0001 1.17275.0001 1.17276.0001 1.17278.0001
1.09194.0001EA 0 GBP
1.09194.0001 1.17275.0001 1.17276.0001 1.17278.0001
Air sampler, MAS-100 NT® Ex
Samplers Air Samplers
The MAS-100 NT® Ex combines all the advantages of the MAS-100 NT® but, in addition, has been tested for use in areas where there are explosive atmospheres.
  • Uses 90 mm petri dishes
  • Programmable delay before sampling starts
  • Pre-selected volumes are individually adjustable
  • Mass flow sensor ensuring a constant airflow
  • Fully automatic calibration cycle with automatic creation of calibration certificate with digital anemometer DA-100 NT® and PC
  • Autoclavable sampling head to avoid any contamination
  • MAS-100 NT EX with HEPA H13 air outlet filter: safety
More Product Information