Potassium dichromate 0.020 mol/l aqueous solution, Titripur® COD_determination, Supelco®

Supplier: Merck
Danger


MQ100

1.09119.1000
1.09119.1000EA 57.1 GBP
1.09119.1000
Potassium dichromate 0.020 mol/l aqueous solution, Titripur® COD_determination, Supelco®
Potassium dichromate

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 3287
Merck Index: 12,07790

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR