Zirconyl chloride octahydrate analytical reagent, Supelco®

Supplier: Merck
Danger

Synonyms: Zirconium(IV) oxide chloride octahydrate, Zirconium dichloride oxide octahydrate, Zirconium(IV) oxychloride octahydrate

M-Clarity™ quality level = MQ300

1.08917.0100
1.08917.0100EA 151 GBP
1.08917.0100
Zirconyl chloride octahydrate analytical reagent, Supelco®
Zirconyl chloride octahydrate
Formula: Cl₂OZr·8H₂O
MW: 322.25 g/mol
Melting Pt: > 150 °C
Density: 1.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149898
CAS Number: 13520-92-8
EINECS: 231-717-9
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 322.28 g/mol
Bulk Density 800 kg/m3
Density 1.91 g/cm3 (20 °C)
pH 1 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR