Hydrogen peroxide 35%, Supelco®

Supplier: Merck
Danger


MQ200

1.08556.9025
1.08556.9025EA 276 GBP
1.08556.9025
Hydrogen peroxide 35%, Supelco®
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8) II
Merck Index: 14,04798

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -24 °C
Boiling Point 110 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Density 1.13 g/cm3 (20 °C)
pH 2 - 4 (H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR