Urea Reag. Ph. Eur., ACS zA, Supelco®

Supplier: Merck

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

MQ200

1.08487.0500 1.08487.1000 1.08487.5000 1.08487.9025
1.08487.0500EA 89.1 GBP
1.08487.0500 1.08487.1000 1.08487.5000 1.08487.9025
Urea Reag. Ph. Eur., ACS zA, Supelco®
Urea
Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 590 g/l (20 °C)
Melting Point 133 °C
Molar Mass 60.06 g/mol
Bulk Density 720 - 760 kg/m3
Vapor Pressure  - 0.1 hPa (20 °C)
Density 1.34 g/cm3 (20 °C)
pH 9 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR