Phenylhydrazinium chloride Reag. Ph. Eur. analytical reagent

Supplier: Merck
Danger

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

M-Clarity™ quality level = MQ200

1.07253.0100 1.07253.0250
1.07253.0100EA 140 GBP
1.07253.0100 1.07253.0250
Phenylhydrazinium chloride Reag. Ph. Eur. analytical reagent
Phenylhydrazinium chloride
Formula: C₆H₉ClN₂
MW: 144.6 g/mol
Melting Pt: 243…246 °C
Storage Temperature: Ambient
MDL Number: MFCD00012924
CAS Number: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: 6.1 II
Merck Index: 12,07447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point 242 - 246 °C (decomposition)
Molar Mass 144.6 g/mol
Bulk Density 330 kg/m3
pH 2.6 - 2.9 (50 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR