Sodium azide tablets for milk preservation, Sigma-Aldrich®
Supplier: Merck
1.06687.0001
1.06687.0001EA 282 GBP
1.06687.0001
Sodium azide tablets for milk preservation, Sigma-Aldrich®
Food and Beverage Test Systems
Simple to use tablet for preservation of milk samples prior to testing. Each 100 mg tablet contains approximately 8 mg of sodium azide.
Packaging: Packed in plastic can.