Sodium pyruvate, Millipore®

Supplier: Merck
Warning

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

MQ100

1.06619.0050 1.06619.0250 1.06619.1000
1.06619.0050EA 49.2 GBP
1.06619.0050 1.06619.0250 1.06619.1000
Sodium pyruvate, Millipore®
Sodium pyruvate
Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR