Sodium nitrate, Supelco®

Supplier: Merck
Warning


MQ200

1.06535.1000 1.06535.9050
1.06535.1000EA 79.3 GBP
1.06535.1000 1.06535.9050
Sodium nitrate, Supelco®
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380
Melting Pt: 308
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1 III
REACH: 01-2119488221-41
Merck Index: 13,08720

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR