Sodium cyanide, Supelco®

Supplier: Merck
Danger

Synonyms: 1-Dodecanesulphonic acid sodium salt

M-Clarity™ quality level = MQ200

1.06437.1000
1.06437.1000EA 74.1 GBP
1.06437.1000
Sodium cyanide, Supelco®
Sodium cyanide
Formula: CNNa
MW: 49.01 g/mol
Boiling Pt: 1496
Melting Pt: 563.9
Density: 1.595 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1 I
Merck Index: 13,08679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR