Sodium bromide, Suprapur® for trace analysis, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

1.06363.0100
1.06363.0100EA 375 GBP
1.06363.0100
Sodium bromide, Suprapur® for trace analysis, Supelco®
Sodium bromide
Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1396
Melting Pt: 753
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now

Specification Test Results

Solubility 905 g/l (20 °C)
Melting Point 755 °C
Molar Mass 102.89 g/mol
Bulk Density 1750 kg/m3
Boiling Point 1393 °C
Vapor Pressure 1.3 hPa (806 °C)
Density 3.20 g/cm3 (20 °C)
pH 5.4 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR