Murexide, Supelco®

Supplier: Merck

Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt

MQ200

1.06161.0005 1.06161.0025
1.06161.0005EA 93 GBP
1.06161.0005 1.06161.0025
Murexide, Supelco®
Murexide
Formula: C₈H₈N₆O₆
MW: 284.19 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6
Merck Index: 13,06330

Order Now

Specification Test Results

Solubility 1 g/l (20 °C)
Molar Mass 284.19 g/mol
Bulk Density 330 kg/m3
pH 5 (1 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR