Dichloromethane 99,8 Stabilised, Suprasolv® hypergrade GC-ECD/FID, Supelco®

Supplier: Merck
Warning

Synonyms: Methylene chloride

MQ100

1.06054.1000 1.06054.2500 1.06054.4000
1.06054.1000EA 99.6 GBP
1.06054.1000 1.06054.2500 1.06054.4000
Dichloromethane 99,8 Stabilised, Suprasolv® hypergrade GC-ECD/FID, Supelco®
Dichloromethane

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionisation Detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥99.9%
Identity (IR) conforms
Evaporation Residue ≤3.0 mg/L
Water ≤0.005%
Colour ≤10 Hazen
GC/ECD (Retention Range 1,2,4-Tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)) ≤2 pg/ml
GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)) ≤2 ng/ml
GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning Area 30 - 600 amu, Individual Signals (n-Tetradecane Standard)) ≤2 ng/ml
Recommended for Analysis of Polychlorinated Dibenzodioxins and Polychlorinated Dibenzofurans (PCDD/F)
This product is stabilised with 2-Methyl-2-Butene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR