Magnesium oxide light Ph. Eur., E530, BP, extra_pure, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

Synonyms: Magnesia usta

1.05862.1000 1.05862.9020
1.05862.1000EA 552 GBP
1.05862.1000 1.05862.9020
Magnesium oxide light Ph. Eur., E530, BP, extra_pure, SAFC®
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/cm³ (25 °C)
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR