Potassium acetate BP, Ph. Eur., E261, extra_pure, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

Synonyms: Acetic acid potassium salt

1.04820.1000 1.04820.5000 1.04820.9025 1.04820.9050
1.04820.1000EA 211 GBP
1.04820.1000 1.04820.5000 1.04820.9025 1.04820.9050
Potassium acetate BP, Ph. Eur., E261, extra_pure, SAFC®
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR