Water, LiChrosolv® UHPLC-MS, Supelco®

Supplier: Merck

Synonyms: Hydrogen oxide

1.03728.1002 1.03728.2002
1.03728.1002EA 49.9 GBP
1.03728.1002 1.03728.2002
Water, LiChrosolv® UHPLC-MS, Supelco®
Water

Highest purity solvents for UHPLC-MS analysis.


  • Suitability tested and specified for UHPLC-MS and UHPLC-UV
  • Specified quality in positive and negative ESI and APCI MS for lowest detection limits and confidence in analyses in all important MS modes - ESI / APCI (+) <2 ppb; ESI / APCI (-) <10 ppb)
  • Lowest impurity profile: For interference-free baselines
  • Lowest levels of trace metal impurities: For minimised metal ion adduct formation (up to <5 ppb)
  • Lowest level of polyethylene glycol (PEG) impurities in our entire UHPLC-MS solvent line up to give you confidence in your results (PEG S/N signal-to-noise ratio <50)


Microfiltration through 0,2 µm filter: Prolonged lifetime of filters and mechanical parts in HPLC systems and reduced risk of column clogging.
Packed in borosilicate glass bottles: Minimises contamination with metal ions.
The borosilicate bottle with GL 45 neck finish can be directly connected to instruments.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Limited Time Sale

Select items below are currently on sale

Order Now

Specification Test Results

Al (Aluminum) ≤ 5 ppb
Ca (Calcium) ≤ 5 ppb
Chloride (Cl) ≤ 10 ppb
Evaporation residue ≤ 0.7 mg/L
Fe (Iron) ≤ 5 ppb
Fluorescence at 254 nm ≤ 0.5 ppb
Fluorescence at 365 nm ≤ 0.5 ppb
K (Potassium) ≤ 5 ppb
Mg (Magnesium) ≤ 10 ppb
Na (Sodium) ≤ 50 ppb
Nitrate (NO3) ≤ 10 ppb
Phosphate (PO4) ≤ 10 ppb
Polyethyleneglycol (PEG) Signal-te-noise ratio ≤ 50
Suitable for LC/MS; lntensity of background mass signals based on reserpine (APCI/ESI negative) ≤ 10 ppb
Suitable for LC/MS; lntensity of background mass signals based on reserpine (APCI/ESI positive) ≤ 2 ppb
Sulphate (SO4) ≤ 10 ppb
Total Organic Carbon ≤ 10 ppb
UHPLC gradient (at 210nm) ≤ 2 mAU
UHPLC gradient (at 254nm) ≤ 0.5 mAU
Filtered by 0.2um filter

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR