Cyclohexane, Uvasol® Spectroscopy, Supelco®

Supplier: Merck
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

MQ100

1.02822.0500 1.02822.2500
1.02822.0500EA 124 GBP
1.02822.0500 1.02822.2500
Cyclohexane, Uvasol® Spectroscopy, Supelco®
Cyclohexane
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 7
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 55 mg/L (20 °C)
Melting Point 6 °C
Molar Mass 84.16 g/mol
Boiling Point 81 °C (1013 hPa)
Vapor Pressure 103 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4264 (20 °C)
Explosion Limit 1.2 - 8.3%(V)
Density 0.78 g/cm³ (20 °C)
pH (H₂O) not applicable
Ignition Point 260 °C
Viscosity Kinematic 1.26 mm²/s (20 °C)
Evaporation Number 3.5
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR