Copper(II) sulphate pentahydrate, EMSURE® Reag. Ph. Eur., ISO analysis_for, Supelco®

Supplier: Merck
Danger

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

MQ300

1.02790.0250 1.02790.1000 1.02790.5000 1.02790.9050
1.02790.0250EA 60.8 GBP
1.02790.0250 1.02790.1000 1.02790.5000 1.02790.9050
Copper(II) sulphate pentahydrate, EMSURE® Reag. Ph. Eur., ISO analysis_for, Supelco®
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Assay (iodometric) 99.0-100.5%
Insoluble matter 0.005% max
Chloride (Cl) 0.0005% max
Total nitrogen (N) 0.001% max
Calcium (Ca) 0.005% max
Iron (Fe) 0.003% max
Potassium (K) 0.001% max
Sodium(Na) 0.005% max
Nickel (Ni) 0.005% max
Lead (Pb) 0.005% max
Zinc (Zn) 0.03% max
Corresponds to ACS,ISO,Reag. Ph Eur

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR