Chloroform, Uvasol® Spectroscopy, Supelco®

Supplier: Merck
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

MQ100

1.02447.0500 1.02447.2500
1.02447.0500EA 80.5 GBP
1.02447.0500 1.02447.2500
Chloroform, Uvasol® Spectroscopy, Supelco®
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 8 g/L (20 °C)
Melting Point -63 °C
Molar Mass 119.38 g/mol
Boiling Point 61 °C (1013 hPa)
Vapor Pressure 211 hPa (20 °C)
Refractive Index 1.45
Density 1.48 g/cm³ (20 °C)
pH (H₂O) no data available
Ignition Point 982 °C
Hazard Class 6.1
Hazard Class Description Toxic materials

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR