tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®

Supplier: Merck
Danger

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

M-Clarity™ quality level = MQ300

1.01849.1000 1.01849.2500 1.01849.9190 1.01849.4000 1.01849.5011
1.01849.1000EA 41.49 GBP
1.01849.1000 1.01849.2500 1.01849.9190 1.01849.4000 1.01849.5011
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
tert-Butyl methyl ether

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications.

Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2
Melting Pt: –108.6 °C
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Limited Time Sale

Select items below are currently on sale

Order Now

Specification Test Results

Purity (GC) ≥ 99,5 %
Identity (IR) conforms
Color ≤ 10 Hazen
Acidity ≤ 0,0005 meq/g
Al (Aluminium) ≤ 0,00005 %
B (Boron) ≤ 0,000002 %
Ba (Barium) ≤ 0,00001 %
Ca (Calcium) ≤ 0,00005 %
Cd (Cadmium) ≤ 0,000005 %
Co (Cobalt) ≤ 0,000002 %
Cr (Chromium) ≤ 0,000002 %
Cu (Copper) ≤ 0,000002 %
Fe (Iron) ≤ 0,00001 %
Mg (Magnesium) ≤ 0,00001 %
Mn (Manganese) ≤ 0,000002 %
Ni (Nickel) ≤ 0,000002 %
Pb (Lead) ≤ 0,00001 %
Sn (Tin) ≤ 0,00001 %
Zn (Zinc) ≤ 0,00001 %
Peroxide (as H₂O₂) ≤ 0,0001 %
Evaporation residue ≤ 0,001 %
Water ≤ 0,03 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR