Nitric acid 69%, EMPARTA® ACS zA, Supelco®

Supplier: Merck
Danger

Synonyms: Nitric acid solution

MQ200

1.01832.2500
1.01832.2500EA 78.9 GBP
1.01832.2500
Nitric acid 69%, EMPARTA® ACS zA, Supelco®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1) II
Merck Index: 13,06610

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -41 °C
Boiling Point 122 °C (1013 hPa)
Vapor Pressure 9.4 hPa (20 °C)
Density 1.41 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive material

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR