Sulphanilic acid ACS, Reag. Ph. Eur. zA, Supelco®

Supplier: Merck
Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid

MQ100

1.00686.0100 1.00686.0250
1.00686.0100EA 62.5 GBP
1.00686.0100 1.00686.0250
Sulphanilic acid ACS, Reag. Ph. Eur. zA, Supelco®
Sulphanilic acid
Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 10 g/l (20 °C)
Melting Point 288 °C decomposes
Molar Mass 173.18 g/mol
Bulk Density 620 kg/m3
Density 1.485 g/cm3 (25 °C)
pH 2.5 (10 g/l, H2O, 20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR